Anti-CYCS Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-833EA 480 EUR
PRSI48-833
Anti-CYCS Mouse Monoclonal Antibody
Antikörper
CYCS antibody can be used in Western Blot, immunohistochemistry starting at 10 μg/mL, and immunoprecipitation.

Protein G Column

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 1% BSA (origin USA), 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CYCS
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR