Anti-NFKB1 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-819EA 480 EUR
PRSI48-819
Anti-NFKB1 Mouse Monoclonal Antibody
NFKB1 antibody can be used in Western Blot, and immunohistochemistry starting at 10 μg/mL.

Affinity Chromatography

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide.

Aliquot and store at 4ºC. Avoid freezing.

Type:
Antigen: NFKB1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR