Anti-PURH Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-813EA 480 EUR
PRSI48-813
Anti-PURH Mouse Monoclonal Antibody
ATIC antibody can be used in ELISA, and immunohistochemistry starting at 10 μg/mL.

Affinity Chromatography

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide at 0.5 mg/ml.

Store ATIC antibody at 4 °C. Do not freeze.

Type:
Antigen: PURH
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Gibbon, Bovine, Dog, Drosophila, Xenopus, Gorilla, Horse, Human, Monkey, Orangutan, Pig, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR