Anti-TIMP2 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-811EA 480 EUR
PRSI48-811
Anti-TIMP2 Mouse Monoclonal Antibody
Antikörper
Metalloproteinase Inhibitor 2 antibody can be used in ELISA, and immunohistochemistry starting at 10 μg/mL.

Affinity Chromatography

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide at 0.5 mg/ml.

Aliquot and store at 4ºC. Avoid freezing.

Type:
Antigen: TIMP2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Gibbon, Bovine, Chicken, Dog, Guinea pig, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR