Anti-HNRNPK Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-810EA 480 EUR
PRSI48-810
Anti-HNRNPK Mouse Monoclonal Antibody
Antikörper
HNRNPK antibody can be used in Western Blot, and immunohistochemistry starting at 5 μg/mL.

Affinity Chromatography

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide at 0.5 mg/ml.

Aliquot and store at 4ºC. Avoid freezing.

Type:
Antigen: HNRNPK
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Dog, Goat, Horse, Human, Monkey, Mouse, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR