Anti-FYN Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-808EA 480 EUR
PRSI48-808
Anti-FYN Mouse Monoclonal Antibody
Antikörper
Protein tyrosine kinase FYN is a non-receptor kinase belonging to the Src subfamily. Various isoforms are produced by alternative splicing.

FYN antibody can be used in Western Blot, immunohistochemistry starting at 1:100, and immunoprecipitation.

Affinity Chromatography

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide at 0.5 mg/ml.

Aliquot and store at 4ºC. Avoid freezing.

Type:
Antigen: FYN
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR