Anti-RPS6KA1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-796EA 480 EUR
PRSI48-796
Anti-RPS6KA1 Rabbit Polyclonal Antibody
Antikörper
RSK1 is an RSK serine/threonine kinase. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

Ribosomal Protein S6 Kinase antibody can be used in ELISA, Western Blot starting at 1:2000, and immunohistochemistry starting at 1:50.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RPS6KA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR