Anti-NFKBIA Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-791EA 480 EUR
PRSI48-791
Anti-NFKBIA Rabbit Polyclonal Antibody
Antikörper
NFKBIA antibody can be used in Western Blot, immunohistochemistry starting at 10 μg/mL, immunocytochemistry, immunoprecipitation, and flow cytometry.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 0.2% gelatin.

Aliquot and store at 4ºC. Avoid freezing.

Type:
Antigen: NFKBIA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR