Anti-MAPK1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-785EA 480 EUR
PRSI48-785
Anti-MAPK1 Rabbit Polyclonal Antibody
Antikörper
ERK2, a MAPK type protein kinase, is a member of the Ras MAPK pathway. This pathway modulates the activity of transcription factors, structural proteins and other protein kinases, leading to biological responses such as cell proliferation and differentiation. Inactive ERK2 associates with MAPK/ERK kinases (MEK) 1 and 2 in the cytosol. Upon stimulation, MEK1/2 phosphorylates and activates ERK2 causing dissociation of the MEK/ERK complex. A portion of the activated ERK2 pool forms dimers and translocates to the nucleus to activate nuclear proteins, such as ELK-1 and MSK1. Cytosolic ERK 2 acts on a different set of substrates, including MNK1/2 and MAPKAP-K1.

MAPK1 antibody can be used in immunohistochemistry starting at 10 μg/mL, immunoprecipitation, and flow cytometry.

Affinity Chromatography

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol, 0.1% gelatin.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MAPK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR