Anti-RPTOR Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-783EA 480 EUR
PRSI48-783
Anti-RPTOR Rabbit Polyclonal Antibody
Antikörper
KIAA1303 antibody can be used in Western Blot, immunohistochemistry starting at 10 μg/mL, immunoprecipitation, and flow cytometry starting at 1:50 - 1:100.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RPTOR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR