Anti-MDM2 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-781EA 480 EUR
PRSI48-781
Anti-MDM2 Mouse Monoclonal Antibody
Antikörper
This product requires antigen retrieval using heat treatment prior to staining of paraffin sections. EDTA pH8. is recommended for this purpose. IHC Control: Breast Carcinoma.

MDM2 antibody can be used in immunohistochemistry starting at 1:50, and flow cytometry starting at 1 µg/1E6 cells.

IgG Fractionation

PBS, pH 7.4, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MDM2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Gibbon, Dog, Human, Monkey, Orangutan, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR