Anti-SLC2A10 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-773EA 480 EUR
PRSI48-773
Anti-SLC2A10 Rabbit Polyclonal Antibody
Antikörper
SLC2A10 antibody can be used in ELISA, and immunohistochemistry starting at 20 μg/mL.

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SLC2A10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gibbon, Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR