Anti-S100 A8 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-769EA 480 EUR
PRSI48-769
Anti-S100 A8 Mouse Monoclonal Antibody
Antikörper
Calgranulin A antibody can be used in Western Blot starting at 1 - 2 μg/mL, and immunohistochemistry starting at 1:50.

Protein G Column

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: S100 A8
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR