Anti-LTA4H Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-750EA 480 EUR
PRSI48-750
Anti-LTA4H Rabbit Polyclonal Antibody
LTA4H antibody can be used in ELISA, and immunohistochemistry starting at 2.5 - 5 μg/mL.

Protein A Column

TBS, 50% glycerol, 0.5 mg/mL BSA, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LTA4H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR