Anti-CAV3 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-742EA 555 EUR
PRSI48-742
Anti-CAV3 Rabbit Polyclonal Antibody
Antikörper
Caveolin proteins are components of caveolae plasma membranes found in most cell types. Caveolin-3 is the muscle-specific form of the caveolin protein family whose members are proposed to be scaffolding proteins for organizing and concentrating certain caveolin-interacting molecules within the caveolae membranes. Mutations in caveolin-3 have been isolated which interfere with caveolin-3 oligomerization and disrupt caveolae formation resulting in limb-girdle muscular dystrophy.

Caveolin-3 antibody can be used in ELISA, and immunohistochemistry starting at 5 μg/mL.

Antiserum

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CAV3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bat, Bovine, Dog, Gibbon, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR