Anti-IL6 Mouse Monoclonal Antibody [clone: [6D9A1,6C12B1]]

Lieferant: ProSci
PRSI48-736EA 619 EUR
PRSI48-736
Anti-IL6 Mouse Monoclonal Antibody [clone: [6D9A1,6C12B1]]
Antikörper
IL-6 antibody can be used in ELISA starting at 1/200 - 1/800, and immunohistochemistry starting at 10 mg/ml.

Ascitic fluid, 0.03% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: IL6
Clonality: Monoclonal
Clone: [6D9A1,6C12B1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR