Anti-INHA Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-732EA 480 EUR
PRSI48-732
Anti-INHA Mouse Monoclonal Antibody
Inhibin alpha antibody can be used in ELISA starting at 1:4000 - 1:20000, Western Blot starting at 1:250 - 1:1500, and immunohistochemistry starting at 5 μg/mL.

Protein A Column

PBS, pH 7.4, 0.03% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: INHA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR