Anti-NFKBIA Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-730EA 0 EUR
PRSI48-730
Anti-NFKBIA Rabbit Polyclonal Antibody
NFKBIA antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 10 μg/mL, and immunofluorescence starting at 10 μg/mL.

PBS, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NFKBIA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR