Anti-EZR Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-716EA 375 EUR
PRSI48-716
Anti-EZR Rabbit Polyclonal Antibody
Antikörper
Ezrin antibody can be used in ELISA, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 1:200 - 1:1000.

PBS, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: EZR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR