Anti-RAF1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-711EA 375 EUR
PRSI48-711
Anti-RAF1 Rabbit Polyclonal Antibody
Antikörper
RAF, a member of the Raf Subfamily of Serine/Threonine Protein Kinases, is part of the RAS-dependent signalling pathway. Once activated, Raf-1 can activate the dual specificity protein kinases MEK1 and MEK2 which in turn activate the serine/threonine specific protein kinases ERK1 and ERK2. Activated ERKs are pleiotropic effectors of cell physiology and play an important role in the control of gene expression involved in the cell division cycle, apoptosis, cell differentiation and cell migration. t(3;8)(p25;q21) translocations involving RAF1 result in mixed parotid gland tumors. Raf1 is an ocogene that has been reported to be overexpressed in many tumor types.

RAF1 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 1:200.

PBS, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR