Anti-H2AFX Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-699EA 514 EUR
PRSI48-699
Anti-H2AFX Rabbit Polyclonal Antibody
H2AFX antibody can be used in ELISA, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 1:200, immunofluorescence starting at 1:200 - 1:1000, and flow cytometry starting at 1:200 - 1:400.

PBS (without Mg2+, Ca2+), pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: H2AFX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR