Anti-GRIA2 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-698EA 480 EUR
PRSI48-698
Anti-GRIA2 Rabbit Polyclonal Antibody
Antikörper
Gria2/Glurb is one of four subunits of the glutamate receptor that is sensitive to alpha-amino-3-hydroxy-5-methyl-4-isoxazolpropionate (AMPA). Glutamate receptors are ligand-activated cation channels that mediate the fast component of excitatory postsynaptic currents in neurons of the central nervous system. These channels are assembled from 4 related subunits, with the Gria2/Glurb subunit rendering the channel almost impermeable to Ca(2+).

GRIA2 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 1:200, and flow cytometry starting at 1:200 - 1:400.

PBS (without Mg2+, Ca2+), pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GRIA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR