Anti-FOXO4 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-696EA 375 EUR
PRSI48-696
Anti-FOXO4 Rabbit Polyclonal Antibody
Antikörper
FOXO4 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 1:200.

Affinity Chromatography

PBS, pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FOXO4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR