Anti-CHEK1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-691EA 375 EUR
PRSI48-691
Anti-CHEK1 Rabbit Polyclonal Antibody
Antikörper
CHK1 prevents entry into mitosis when damaged or unligated DNA is detected by phosphorylating cdc25a, cdc25b, and cdc25c. Phosphorylation of cdc25c creates a binding site for protein 14-3-3, which inhibits cdc25c and prevents activation of the cdc2-cyclin b complex.

CHEK1 antibody can be used in ELISA, Western Blot starting at 1:200 - 1:1000, immunohistochemistry starting at 1:200, immunohistochemistry starting at 1:200 - 1:1000, and immunofluorescence starting at 1:200 - 1:1000.

PBS, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CHEK1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR