Anti-TYMS Sheep Polyclonal Antibody

Lieferant: ProSci
PRSI48-685EA 480 EUR
PRSI48-685
Anti-TYMS Sheep Polyclonal Antibody
Antikörper
TYMS antibody can be used in ELISA, and immunohistochemistry starting at 10 μg/mL.

IgG Fractionation

0.02 M Potassium Phosphate, 0.15 M NaCl, pH 7.2 with 0.01% (w/v) Sodium Azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TYMS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR