Anti-LEP Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-684EA 480 EUR
PRSI48-684
Anti-LEP Rabbit Polyclonal Antibody
Antikörper
Leptin has been implicated in the control of appetite and metabolism.

Leptin antibody can be used in ELISA, and immunohistochemistry starting at 40 μg/mL.

Antiserum

Contains 0.01% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LEP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR