Anti-LDB2 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-678EA 480 EUR
PRSI48-678
Anti-LDB2 Rabbit Polyclonal Antibody
Antikörper
LDB2 (LIM homeobox protein cofactor CLIM-1a). It is suggested that LIM domain binding proteins (LDB1, LDB2 and LDB3) act synergistically to enhance transcriptional efficiency by acting as co-factors for LIM homeodomain and Otx class transcription factors both of which have essential roles in development. LDB2 is a close homologue of LDB1. LDBs are homologs of the Drosophila Chip protein. Three members of the ldb gene family (Ldb1, Ldb2 and Ldb3) from the zebrafish, Danio rerio, share 95%, 73% and 62% amino acid identity with mouse Ldb1, respectively.

LDB2 antibody can be used in ELISA starting at 1:3000 - 1:12000, Western Blot starting at 1:2000 - 1:10000, and immunohistochemistry starting at 2.5 - 5 μg/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Store LDB2 antibody at -20 °C prior to opening. Dilute only prior to immediate use. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LDB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Chicken, Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR