Anti-MRE11A Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-672EA 480 EUR
PRSI48-672
Anti-MRE11A Rabbit Polyclonal Antibody
Antikörper
Mre11 is a component of the MRN complex (Mre11/Rad50/Nbs1), which plays a central role in double-strand break (DSB) repair, DNA recombination, maintenance of telomere integrity and meiosis. The complex possesses single-strand endonuclease activity and double-strand-specific 3'-5' exonuclease activity, which are provided by MRE11A. RAD50 may be required to bind DNA ends and hold them in close proximity. This could facilitate searches for short or long regions of sequence homology in the recombining DNA templates, and may also stimulate the activity of DNA ligases and/or restrict the nuclease activity of MRE11A to prevent nucleolytic degradation past a given point. The complex may also be required for DNA damage signaling via activation of the ATM kinase. In telomeres the MRN complex may modulate t-loop formation. Mre11 is a nuclear protein that is widely expressed and localizes to discrete nuclear foci after treatment with genotoxic agents (including irradiation). Alternative splice isoforms are recorded for this protein.

MRE11A antibody can be used in ELISA starting at 1:5000 - 1:25000, Western Blot starting at 1:500 - 1:3000, immunohistochemistry starting at 2.5 μg/mL, and immunofluorescence.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MRE11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR