Anti-KALRN Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-649EA 480 EUR
PRSI48-649
Anti-KALRN Rabbit Polyclonal Antibody
Antikörper
Duet is a TRIO/TRAD serine/threonine kinase with Dbl-Homology (DH) and Pleckstrin-Homology (PH) domains. Duet protein was localized to actin-associated cytoskeletal elements, suggesting a role in the cytoskeleton-dependent cell function. Huntington's disease is an inherited neurodegenerative disease which occurs when the widely expressed protein huntingtin contains an expanded glutamine repeat. HAPIP protein binds to HAP1, which is a huntington binding protein that may function in vesicle trafficking.

KALRN antibody can be used in immunohistochemistry starting at 20 μg/mL.

PBS, pH 7.4, 0.09% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: KALRN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR