Anti-RNF34 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-642EA 480 EUR
PRSI48-642
Anti-RNF34 Rabbit Polyclonal Antibody
Antikörper
RNF34, also known as RING finger protein 34, RING finger protein RIFF, FYVE-RING finger protein Momo, Human RING finger homologous to inhibitor of apoptosis protein, hRFI, Caspases-8 and -10-associated RING finger protein 1, CARP-1 and Caspase regulator CARP1, is a novel modulator of NF-κB activation. RNF34 possesses E3 ubiquitin protein ligase activity and has been shown to regulate the levels of CASP8 and CASP10 by targeting them for proteasomal degradation. RNF34 protects cells against apoptosis induced by TNF and may bind phosphatidylinositol phosphates. RNF34 is localized to both the nucleus and the cytoplasm. In the nucleus, a distinct speckled pattern is observed by immunofluorescence microscopy. In the cytoplasm, the protein is associated with membranes. RNF34 is expressed ubiquitously and can be detected in heart, brain, liver, skeletal muscle, kidney, pancreas, spleen, thymus, prostate, testis, ovary, colon and leukocytes. Multiple transcript variants have been reported for this protein.

RNF34 antibody can be used in ELISA starting at 10 - 20 μg/mL, Western Blot starting at 1 - 2 μg/mL, and immunohistochemistry starting at 5 μg/mL.

Protein A Column

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RNF34
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR