Anti-TBK1 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-639EA 480 EUR
PRSI48-639
Anti-TBK1 Mouse Monoclonal Antibody
Antikörper
TBK1, also known as NF-kappa-B-activating kinase (NAK), is an IKAPPAB type protein kinase. TBK1 protein contains a N-terminal kinase domain and two putative coiled-coil regions in the C-terminal region. It has been shown to associate with TANK and TRAF2 in a TBK1-TANK-TRAF2 signaling complex, which is believed to function upstream of the IKK complex in the TRAF-mediated activation of NF-kappaB.

TBK1 antibody can be used in ELISA starting at 1:10000, and immunohistochemistry starting at 1:200 - 1:1000.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TBK1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Gibbon, Bovine, Dog, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR