Anti-NFYA Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-636EA 505 EUR
PRSI48-636
Anti-NFYA Rabbit Polyclonal Antibody
Antikörper
NFYA antibody can be used in immunohistochemistry starting at 10 μg/mL.

Sterile Filtration

Serum - liquid, 0.01% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NFYA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR