Anti-PMP22 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-619EA 530 EUR
PRSI48-619
Anti-PMP22 Rabbit Polyclonal Antibody
PMP22 is a major component of myelin expressed in the compact portion of essentially all myelinated fibers in the peripheral nervous system and is produced predominantly by Schwann cells. Studies in injured nerve suggested a role during Schwann cell growth and differentiation. PMP22 is expressed in cranial nerves but not in the mature central nervous system. Studies suggest a possible role of the PMP22 gene in the development of neoplasia in patients with neurofibromatosis and in the myelin degenerative Charcot-Marie-Tooth disease linked to chromosome 17p.

PMP22 antibody can be used in ELISA, and immunohistochemistry starting at 5 μg/mL.

Affinity Chromatography

PBS, pH 7.4, 0.2% BSA, 0.05% sodium azide.

Aliquot and store at 4ºC. Avoid freezing.

Type:
Antigen: PMP22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR