Anti-NEK2 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-615EA 480 EUR
PRSI48-615
Anti-NEK2 Rabbit Polyclonal Antibody
Antikörper
NEK2, also called serine/threonine-protein kinase Nek2NimA-related protein kinase 2, NimA-like protein kinase 1 and HSPK 21, is a protein kinase involved in mitotic regulation that may have a role at the G2-M transition and may also play a role in meiosis. NEK2 has a nuclear localization and accumulates throughout S phase and shows maximal levels in late G2. This expression pattern is highly reminiscent of that of A and B cyclins.

NEK2 antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 10 μg/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Store NEK2 antibody at -20 °C prior to opening. Dilute only prior to immediate use. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NEK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR