Anti-CBX4 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-604EA 505 EUR
PRSI48-604
Anti-CBX4 Rabbit Polyclonal Antibody
PC2 is the human homolog of the Drosophila 'Polycomb' (Pc) protein which has been identified as a gene family member associated with a cellular memory system that is responsible for the inheritance of gene activity by progeny cells. The human Pc homolog gene is more closely related to a Xenopus Pc homolog, XPc, than to a previously described human Pc homolog, CBX2 (hPc1). However, the hPc2 and CBX2/hPc1 proteins are shown to colocalize in interphase nuclei of human U-2 OS osteosarcoma cells, suggesting that the proteins are part of a common protein complex. The human protein is believed to function as a repressor of proto-oncogene activity and that interference with hPc2 function can lead to derepression of proto-oncogene transcription and subsequently to cellular transformation. Other reports describe PC2 as a protein that has SUMO E3 activity for the corepressors CtBP and CtBP2.

CBX4 antibody can be used in Western Blot, and immunohistochemistry starting at 20 μg/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CBX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR