Anti-SOCS4 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-585EA 530 EUR
PRSI48-585
Anti-SOCS4 Rabbit Polyclonal Antibody
SOCS4 antibody can be used in ELISA starting at 1:10000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 1:200 - 1:1000, and immunofluorescence starting at 1:200 - 1:1000.

Affinity Chromatography

PBS, pH 7.4, 0.05% sodium azide.

Aliquot and store at 4ºC. Avoid freezing.

Type:
Antigen: SOCS4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR