Anti-RGS2 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-550EA 480 EUR
PRSI48-550
Anti-RGS2 Rabbit Polyclonal Antibody
Antikörper
Regulators of G-protein signaling (RGSs) are a protein family containing a conserved 120 amino acid domain (RGS domain), which bind directly to activated G-alpha subunits (Gai and Gaq) and act as GTPase-activating proteins (GAPs). One of these proteins, RGS2,is expressed in thyroid, heart. thyroid, and monocyte-derived DCs. The function of RGS2 as a regulator of Gq is also dependent on regulatory sequences in its amino-terminal domain that direct its localization to the plasma membrane. Recent studies have shown that RGS2 is critical for blood pressure homeostasis, and has been suggested to serve as an effector of the NO-cGMP pathway that promotes vascular relaxation based on studies of aortic rings in vitro.

RGS2 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 10 μg/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RGS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR