Anti-MAGEA1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-549EA 375 EUR
PRSI48-549
Anti-MAGEA1 Rabbit Polyclonal Antibody
Antikörper
MAGE was originally isolated from human melanoma cells as an antigen that is recognizes by cytolytic T lymphocytes. It has since been documented in other carcinomas. In addition, expression has been documented in highly proliferating germ cells.

MAGEA1 antibody can be used in ELISA, and immunohistochemistry starting at 20 μg/mL.

Purified

PBS, pH 7.4, 0.05% sodium azide.

Aliquot and store at 4ºC. Avoid freezing.

Type:
Antigen: MAGEA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR