Anti-LMNA Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-519EA 480 EUR
PRSI48-519
Anti-LMNA Rabbit Polyclonal Antibody
Antikörper
Lamin A and lamin C are alternative splicing products of the lamin A/C gene that is responsible for autosomal dominant Emery-Dreifuss muscular dystrophy (AD-EDMD). Aberrant expression patterns of nuclear lamins have been described in various types of cancer depending on the subtype of cancer, its aggressiveness, proliferative capacity and degree of differentiation. In general, the expression of A-type lamins (lamins A and C) has been correlated with a non-proliferating, differentiated state of cells and tissues. Lamins A and C, the products of the LMNA gene, are nuclear intermediate filament proteins and are the major structural components of the lamina network that underlies and supports the nuclear envelope.

Lamin A antibody can be used in ELISA, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 2.5 μg/mL.

Antiserum

PBS containing 0.05% BSA and 0.05% sodium azide

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LMNA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gibbon, Dog, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR