Anti-RPS6KB1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-518EA 375 EUR
PRSI48-518
Anti-RPS6KB1 Rabbit Polyclonal Antibody
Antikörper
Ribosomal Protein S6 Kinase (P70-S6K) p70 RSK serine/threonine subfamily of protein kinases.

Ribosomal Protein S6 Kinase antibody can be used in immunohistochemistry starting at 20 μg/mL, and flow cytometry.

PBS, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RPS6KB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR