Anti-CDK1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-513EA 375 EUR
PRSI48-513
Anti-CDK1 Rabbit Polyclonal Antibody
Antikörper
CDC2/CDK1, a CDC2/CDK -type protein kinase, regulates cell cycle transitions G1 to S and G2 to M. CDC2/CDK1 is maintained in an inactive state by phosphorylation by kinases MYT1 and WEE1. The active kinase, called the M-phase promoting factor, consists of a heterodimer with cyclin B, and final activation at G2/M results from CDC25 phosphatase-mediated dephosphorylation of CDC2/CDK1. The drug taxol has been shown to activate CDC2/CDK1 in MDA-MB-435 breast cancer cells, leading to cell cycle arrest at G2/M and to apoptosis. CDC2 has also been implicated in neurofibrillary degeneration in Alzheimer's disease and other neurodegenerative diseases.

CDC2 antibody can be used in Western Blot, immunohistochemistry starting at 20 μg/mL, and flow cytometry.

PBS, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CDK1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR