Anti-CAMK2A Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-510EA 375 EUR
PRSI48-510
Anti-CAMK2A Rabbit Polyclonal Antibody
Antikörper
CAMK2Alpha, together with the beta, delta, and gamma subunits, constititute the CAMK2 heteromultimeric holoenzyme serine/threonine kinase. CAMK2A has been implicated in learning, where it potentiates synaptic transmission by phosphorylating AMPA receptors. Variants are produced by alternative splicing.

CAM Kinase 2 alpha antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunoprecipitation

PBS (without Mg2+, Ca2+), pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CAMK2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR