Anti-FAM129A Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-509EA 375 EUR
PRSI48-509
Anti-FAM129A Rabbit Polyclonal Antibody
Antikörper
FAM129A antibody can be used in Western Blot, immunohistochemistry starting at 20 μg/mL, and flow cytometry.

PBS, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FAM129A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR