Anti-BRCA1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-508EA 480 EUR
PRSI48-508
Anti-BRCA1 Rabbit Polyclonal Antibody
Antikörper
Associated with several familial cancers, including breast cancer, ovarian cancer, and prostate cancer. The cumulative lifetime risk of breast cancer for women who carry mutations in BRCA1 is approximately 92%, while the cumulative lifetime risk for noncarriers is estimated to be 10%. BRCA1 and BRCA2 participate together in a pathway (or pathways) associated with the activation of double-strand break repair and/or homologous recombination.

BRCA1 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

PBS, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: BRCA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR