Anti-BCL2 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-505EA 375 EUR
PRSI48-505
Anti-BCL2 Rabbit Polyclonal Antibody
Antikörper
Bcl-2 inhibits apoptosis. Translocations that lead to overexpression of this gene are associated with cancer (lymphoma in the case of translocation to the Ig heavy chain locus).

BCL2 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

PBS, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: BCL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR