Anti-NRG3 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-463EA 375 EUR
PRSI48-463
Anti-NRG3 Rabbit Polyclonal Antibody
Antikörper
Neuregulin-3 antibody can be used in ELISA starting at 1:1000 - 1:5000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 1:500, and immunoprecipitation

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NRG3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR