Anti-LRP2 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-441EA 375 EUR
PRSI48-441
Anti-LRP2 Rabbit Polyclonal Antibody
Antikörper
LRP2 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 1:500, immunofluorescence, and immunoprecipitation.

Protein G Column

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LRP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR