Anti-OAS1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-425EA 375 EUR
PRSI48-425
Anti-OAS1 Rabbit Polyclonal Antibody
Antikörper
OAS1 antibody can be used in ELISA starting at 1:10000 - 1:50000, Western Blot starting at 1:5000 - 1:10000, immunohistochemistry starting at 2.5 μg/mL, and immunoprecipitation

Protein G Column

PBS, 0.09% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: OAS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR