Anti-PTGER3 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-422EA 619 EUR
PRSI48-422
Anti-PTGER3 Rabbit Polyclonal Antibody
Antikörper
Prostaglandin E Receptor EP3 is a member of the Prostanoid Receptor subfamily. Along with several other receptor subtypes (EP1, EP2, and EP4), it mediates the diverse biological effects of Prostaglandin E2 (PGE2). PGE2 has been implicated in numerous processes, including immune response modulation, male erectile function, induction of labor, cervical cancer, control of blood pressure, and asthma. Various isoforms are produced by alternative splicing.

PTGER3 antibody can be used in immunohistochemistry starting at 10 μg/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PTGER3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Gorilla, Horse, Human, Monkey, Mouse, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR