Anti-GPR18 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-420EA 480 EUR
PRSI48-420
Anti-GPR18 Rabbit Polyclonal Antibody
Antikörper
GPR18 is an Orphan-A GPCR with no known ligand.

GPR18 antibody can be used in ELISA starting at 1:10000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 1:200 - 1:400, and immunofluorescence starting at 1:200 - 1:1000.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPR18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Gorilla, Horse, Human, Monkey, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR